Learn More
Innovating Science™ Soaps and Detergents
Examine similarities and differences in soaps and detergents.
Supplier: Innovating Science™ IS9351
Description
- Look at the properties of soap — hand dishwashing detergent and machine dishwashing detergent; both are provided in the kit
- Activities include testing pH, examining the effect of soap and detergent on the surface tension of water, foaming ability, fat emulsification and the performance of soap and detergent in hard water
Order Info
Class size: 15 groups.
Specifications
Soaps and Detergents Kit | |
7 to 14 | |
To examine similarities and differences in soaps and detergents | |
1 | |
15 Lab Groups |
White | |
13 to 20 | |
1 x 25mL Liquid soap, 1 x 25mL dishwashing detergent (hand), 1 x 25mL dishwashing detergent (machine), 1 x 60mL vegetable oil, 1 x 60mL Calcium Chloride, 5% solution, 60 disposable medicine cups, 60 Polystyrene test tubes, 60 capillary tubes, 60 graduated | |
12 x 9.25 x 4 in. |