Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Innovating Science™ Molarity Lab Investigation Educational Kit
SCI_ED

Designed to introduce students to the concept of molarity. Innovating Science Molarity Lab Investigation Educational Kit helps students to prepare a solution with a specific molarity demonstrating the importance of good laboratory technique.

Supplier:  Innovating Science™ IS2519

Catalog No. S28491


Add to Cart

Alert:

WARNING: This product can expose you to chemicals including Phenolphthalein, which is known to the State of California to cause cancer.


Description

Description

  • Students first try to make a solution with a specific molarity demonstrating the importance of good laboratory technique
  • Then, students perform a titration on a known acid solution before using the concepts they have learned to identify the concentrations in three unknown solutions
  • Contains enough materials for 15 groups
  • Teacher's manual and student Study Guide copymasters are included
  • Kit Includes:
    • 0.1 M Sodium Hydroxide (5 L)
    • 0.3 M Hydrochloric Acid (400 mL)
    • Phenolphthalein Solution (45 mL)
    • Sucrose (1200 g)
    • Unknown Hydrochloric Acid #1 (400 mL)
    • Unknown Hydrochloric Acid #2 (400 mL)
    • Unknown Hydrochloric Acid #3 (400 mL)
Specifications

Specifications

Molarity Lab Investigation Educational Kit
7 to 14
Educational use
12 x 12 x 12 in.
White
13 to 20
1 Kit
15 Lab Groups
Product Suggestions

Product Suggestions

SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit