Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Innovating Science™ Lab Pillows
SCI_ED

Absorbent lab pillows designed to absorb almost any liquid to safely handle lab spills. Pack of 18 pillows: 4 x 14 x 1 in.

Supplier:  Aldon Corporation ADKK8510KIT

Catalog No. S28617


Add to cart

Description

Description

Each 4 x 14 x 1 in. lab pillow absorbs 0.22 gal (750mL) of solvent. Not to be used on hydrofluoric acid.
Specifications

Specifications

White
750 mL
Absorbing liquid laboratory spills
4 x 14 x 1 in.
0.22 gal.
Spill Control Pillow
18/Pk.
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit