Learn More
Innovating Science™ EZ Prep Chemicals (A-M)
SCI_ED
Capsules are pre-measured to make a chemical solution.
Supplier:  Aldon Corporation IS4062
 View more versions of this product
Catalog No. S4062A
Add to Cart
Description
Description
  • Add to distilled water and stir to make the amount of solution indicated
  • There is no need to handle chemicals as the chemicals are contained within the capsule
  • Decrease preparation time since no weighing of chemicals is required
  • Decrease disposal costs because no extra chemicals remain
Specifications
Specifications
7 Lab
Bromocresol Green 0.4%
White
Preparations of chemical solutions
Capsule
1 Capsule
English
Chemistry
Product Suggestions
Product Suggestions