Learn More
Innovating Science™ EZ Prep Chemicals (A-M)
Capsules are pre-measured to make a chemical solution.
Supplier: Aldon Corporation IS4057
Description
- Add to distilled water and stir to make the amount of solution indicated
- There is no need to handle chemicals as the chemicals are contained within the capsule
- Decrease preparation time since no weighing of chemicals is required
- Decrease disposal costs because no extra chemicals remain
Specifications
7 Lab | |
Bromothymol Blue 0.1% | |
White | |
Preparations of chemical solutions |
Capsule | |
1 Capsule | |
English | |
Chemistry |