Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Novus Biologicals™ InnovaCoat (R) GOLD 20nm Mini Antibody Labeling Kit
Use simple pipetting operations to covalently attach antibodies, proteins and other biomolecules to ultra-stable InnovaCoat GOLD nanoparticles. Hands-on time is about two minutes and the conjugate is ready within fifteen.
Supplier: Novus Biologicals™ 2290010
Alert:
For Research Use Only
Description
Features- Proprietary surface coat reduces risk of sub-optimal antibody performance
- Ultra-stable
- Covalently link antibodies and analytes
- Simple conjugation processes
- Different surface chemistries
- Antibody-GOLD conjugates in just 15 minutes
- Enhanced assay sensitivity
- Protective coat prevents metal-antibody interactions
- Coating procedure not dependent on isoelectric point of antibody
- Rapid screening of multiple antibodies for assay development
- Reduced antibody loading without aggregation
- No pH titrations
- Fully scalable – from R&D to production
- Custom InnovaCoat™ labeling services
Specifications
Store at 4°C. Do not freeze. | |
10 Tests | |
Antibody Labeling Kit |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction