Learn More
Perkin Elmer US LLC 2.0 mm I.D. Sapphire Injector for Optima 2x00/4x00/5x00/7x00 DV
Supplier: Perkin Elmer US LLC N0775247
This Sapphire Injector is for use with Optima 2.0 mm I.D. Sapphire Injector for Optima 2x00/4x00/5x00/7x00 DV ICP-OES series instruments. Sapphire injectors are ideal for silicon applications.