Learn More
Perkin Elmer US LLC Injector Lock Collar for Avio 200/500
Supplier: Perkin Elmer US LLC N0791163
![Encompass_Preferred](/content/dam/fishersci/glyphs/encompass_preferred.png)
This lock collar is included in the Cyclonic Injector Adapter Kit for Avio 200/500 (N0790141) and the Scott Injector Adaptor Kit for Avio 200/500 (N0790156).