Learn More
Clontech Labs 3P In-Fusion Snap Assembly Master Mix | 250 Rxns
![SDP](/content/dam/fishersci/glyphs/sdp.png)
Supplier: Clontech Labs 3P 638949
![Encompass](/content/dam/fishersci/glyphs/encompass.png)
In-Fusion Snap Assembly Master Mix is designed for fast, directional cloning of one or more fragments of DNA into any vector. This proprietary master mix fuses DNA fragments (e.g., PCR-generated sequences and linearized vectors) efficiently and precisely by recognizing a 15-bp overlap at their ends. This 15-bp overlap can be engineered into the primers designed for PCR amplification of the desired sequences. In Fusion Snap Assembly Master Mix offers high efficiency, even for applications that can be challenging, including the cloning of long fragments, short oligonucleotides, and multiple fragments.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.