Learn More
Perkin Elmer US LLC Infrared Window for Microscopic FTIR Analysis - BaF2, 1 mm
Supplier: Perkin Elmer US LLC N9302611
Infrared windows allow for support of microsamples for infrared analysis with microscopes, such as PerkinElmer's Spotlight Systems. These windows are available in the following infrared crystals: BaF2, KBr, NaCl, and ZnSe.