Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Influenza A H9N2 Hemagglutinin anti-Virus, HRP, Antibody Pair, Novus Biologicals™
Antibody pair for Solid phase sandwich ELISA
$1112.50
Specifications
Antigen | Influenza A H9N2 Hemagglutinin |
---|---|
Applications | ELISA |
Conjugate | HRP |
Regulatory Status | RUO |
Target Species | Virus |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB183326
|
Novus Biologicals
NBP3067055PLATES |
5 Plates |
Each for $352.89
|
N/A | |||||
Due to product restrictions, please Sign In to purchase or view availability for this product. | |||||||||
NB183325
|
Novus Biologicals
NBP30670515PLATES |
15 Plates |
Each for $1,112.50
|
|
|||||
Description
Solid phase sandwich ELISA for quantitative determination of Influenza A H9N2 HemagglutininSpecifications
Influenza A H9N2 Hemagglutinin | |
HRP | |
Virus | |
Storage of components varies. See protocol for specific instructions. |
ELISA | |
RUO | |
HA, Hemagglutinin, Hemagglutinin HA1 chain, Hemagglutinin HA2 chain | |
Antibody Pairs |
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title