Learn More
SPEX CERTIPREP LLC 3P INFLUENT TSS STANDARD
Supplier: SPEX CERTIPREP LLC 3P QCI055
(NSI by Zeptometrix) QC all of your solids analyses with our ready-to-use, NIST traceable solids standards. Stable and remarkably easy to use, simply shake well and pour an appropriate aliquot. No pipeting, no dilutions minimize error. Packaged in an economical 4x1 liter case yielding up to nearly 250 QC analyses for the high level concentrations. Certifi cate includes true values, acceptance limits and NIST traceability information.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.