Learn More
Electron Microscopy Sciences INFINITY1 3.1 Megapixel USB 2.0 Microscopy Camera, CMOS, Color
SDP
Supplier:  Electron Microscopy Sciences 95102
Encompass_Preferred
INFINITY1 CMOS USB 2.0 digital cameras are designed to be a cost-effective, versatile solution for clinical, life science, materials science and educational professionals. Features include auto white balance, full exposure control, programmable gain, sub-windowing, and region of interest. It is an excellent fit for documentation and archiving applications.
Features 2, 3, and 5 mexapixel resolution. It is perfect for documentation and archiving applications and has fast frame rates.
Sensor: CMOS
Type: Color
Catalog No. 50-323-09