Learn More
Seca Corp 416 Infantometer length measuring for babies and toddlers, centimeters-only
Supplier:  Seca Corp 4161721009
Encompass_Preferred
The sturdy construction and stability of the infantometer makes it quick and easy to measure babies and toddlers up to two years old. The board is generously designed and the raised and softly rounded sides guide the baby safely into the right position. Permanently mounted with rollers on two guide rails, the footpiece slides smoothly along the scale. Since the footpiece is lockable, the baby can be removed and the result noted later. • Robust, extra-large construction for precise measuring • Ensures correct lying position due to trough-shaped measuring surface • Measurement scale printed along top side for easy reading • Space-saving • Measuring range: 33 – 100 cm • Graduation: 1 mm
Catalog No. NC1961655