Learn More
Yamato Scientific America, Inc. Yamato 318L Incubator with window, General Purpose, 115V
Supplier: Yamato Scientific America, Inc. IC803CW
Economical general purpose 318L natural convection incubator with observation window. Dual door system permits contents to be viewed easily without disrupting atmosphere of the incubator. Easy to use digital setting display and timer. Air jacket technology ensures even and efficient heat distribution throughout the chamber. Standard equipped with various functions such as self-diagnostic, calibration off-set, overheat prevention and key lock. Operating temperature range: +5~ 80°C
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.