Learn More
Advansta Inc Incubation Tray, opaque black body and lid, 6x9 cm, 4/pack
Supplier:  Advansta Inc L07039004
Encompass
These lidded incubation trays have a perfectly smooth interior surface, making them ideal for staining and washing electrophoresis gels and membranes. Trays are manufactured to prevent adsorption of antibodies.
Catalog No. NC1638170