Learn More
Electron Microscopy Sciences CoverWell™ Incubation Chambers, 50 mcL, 0.5 mm depth
Supplier: Electron Microscopy Sciences 7032405
CoverWell incubation reusable chambers are reusable, easy to apply incubation chambers that attach to slides without the use of adhesive.
Microwavable for permeabilization and retrieval steps.
Preserve kinetic (non-capillary) fluid dynamics for better reagent mixing and lower backgrounds.
Eliminate precipitate deposits on specimens by incubating slides and specimens upside down during enzymatic color precipitation reaction.
Provides an exceptionally secure seal during submerged water bath and/or high temperature incubations.
Rnase and Dnase free, hydrophobic
Easily removed (reusable)
Adheres to wet or dry surfaces.
Volume: 50 µl
Depth: 0.5 mm
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.