Learn More
Invivogen INCOMP FREUNDS ADJUVANT 6X10ML
SDP
Supplier:  Invivogen VACIFA60
Encompass
FA (Incomplete Freunds adjuvant) is one of the most commonly used adjuvants in research. It is prepared from non-metabolizable oils.IFA can be used to produce water-in-oil emulsions of antigens.IFA induces a predominantly Th2-biased response through the formation of a depot at the injection site and the stimulation of antibody producing plasma cells. INVIVOGEN mfg# VAC-IFA-60
Catalog No. NC2020223
Explore available promotions