Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Inclined Plane Board
![SCI_ED](/content/dam/fishersci/glyphs/scied.png)
Durable and consistent.
Supplier: Christensen Educational Materials 9121
This item is not returnable.
View return policy
Description
Extruded aluminum maintains rigidity for reliable performance.
- Made of 0.38 in. (10mm) thick Masonite™
- 1.5 in. dia. (3.8cm) cone-bearing plastic pulley mounted on one end Dimensions: 5.3 × 0.63 × 49 in. (13.3 × 1.6 × 122cm)
- 0.6 in. (1.5cm) guide rails resist warping and act as a guide for a Hall's carriage and coefficient of friction blocks
Specifications
Inclined Plane Board | |
Act as guide for halls carriage | |
3.8cm dia. Plastic pulley | |
48.03 in. | |
5.23 in. |
Masonite Board | |
Halls carriage | |
48.03 x 5.23 in. (122 x 13.3 cm) | |
122 cm | |
13.3 cm |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction