Learn More
MilliporeSigma™ Supelco™ In-Line Pressure Regulators
Install downstream of a gas cylinder regulator to adjust pressure closer to the point of use
Supplier: MilliporeSigma™ Supelco™ 23882
Description
- Adjusts pressure closer to the point of use
- Includes 1/8 in. Swagelok Nuts and Ferrules
- Maximum Input Pressure: 400 psi
Specifications
Air, H2, N2, Ar, He | |
0-100 psi (Outlet) | |
In-Line Pressure Regulator |
1/8 in. Swagelok Nuts and Ferrules | |
High Purity Version |