Learn More
American International Electric IMPULSE SEALER 8IN
Supplier: American International Electric AIE200
IMPULSE HAND SEALERS,Seal Length:8 in. ,Seal Thickness:6 mil,Seal Width:2 mm,Watts:260 W,Weigh t :8 lbs