Learn More
Ace Glass, Inc. Impinger, 25mL, complete with 12/5 socket inlet, 12/5 ball outlet stopper, calibrated nozzle
SDP
Supplier:  Ace Glass, Inc. 753129
Encompass_Preferred
25mL midget impinger used for sampling small air volumes at low velocities. Includes 12/5 socket inlet and 12/5 ball outlet stopper with calibrated nozzle that delivers 0.09-0.11 CFM at 30.5cm vacuum.
Catalog No. 50-979-318