Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Impacto™ Impactosol Flat GEL Insoles
Impacto™ Impactosol Flat Gel Insoles help relieve foot fatigue and are ideal for long term wear
Supplier: Impacto™ IMFLATD
Description
Impacto™ IMPACTOSOL Anti-Fatigue Insoles
- Made with VEP gel with Poron foam top layer provides cushioning and even distribution of weight to protect pressure points
- Anti-bacterial
- For use with most types of footwear including low cut footwear Can be trimmed for a perfect fit
Specifications
Blue | |
22.86 cm | |
Men: 11 to 13, Women: 13 to 14 | |
Unisex |
9 in. | |
Gel | |
Open cell structure full flat insoles | |
Impactosol Insoles |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction