Learn More
Impacto™ VEP Soft padded Knee Pads
Impacto™ 803-20 VEP padded soft Kneepads provides protection from cumulative trauma
Supplier: Impacto™ 80320110000
Description
Impacto™ Soft Padded Kneepads
- Impact absorbing layered VEP and foam padding with grain leather cover
- Upper and lower elastic straps with hook and loop closure
- Protection from cumulative trauma
- Impact, cushioning and abrasion protection
Specifications
Hook and loop | |
Stretch polyester and grain leather; Visco elastic polymer (VEP) padding | |
8 in. | |
Polyester |
Knee Pad | |
20.32 cm | |
Universal |