Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Thermo Scientific™ ImmunoCAP™ Specific IgG4 Reagents for Phadia™ 1000/250
$1500.00
Specifications
For Use With (Equipment) | Phadia™ 1000/250 |
---|---|
Content And Storage | 2°C to 8°C |
Catalog Number | Mfr. No. | Quantity | Product Type | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Product Type | Price | Quantity & Availability | ||||
10-948-102P
|
Thermo Scientific™
10948102 |
2 Vials, 100 Detections | Specific IgG4 Conjugate 50 |
Pack of 100 for $1,500.00
|
|
||||
10-948-202P
|
Thermo Scientific™
10948202 |
1 Strip, 1 Curve | Specific IgG4 calibrator strip |
Item Discontinued
This item has been discontinued by the supplier. Please Sign In to view product availability in your area. View Alternate Product |
N/A | ||||
10-948-302P
|
Thermo Scientific™
10948302 |
1 Strip, 3 Runs | Specific IgG4 curve control strip |
Item Discontinued
This item has been discontinued by the supplier. Please Sign In to view product availability in your area. View Alternate Product |
N/A | ||||
Description
Specifications
Phadia™ 1000/250 | |
2°C to 8°C |
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title