Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Thermo Scientific™ ImmunoCAP™ Specific IgE Reagents for Phadia™ 5000/2500/1000/250
Provide an objective measurement of the circulating IgE antibodies and the sensitization to a specific allergen
$1602.00 - $6408.00
Catalog Number | Mfr. No. | For Use With (Equipment) | Quantity | Product Type | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | For Use With (Equipment) | Quantity | Product Type | Price | Quantity & Availability | |||
10-936-001P
|
Thermo Scientific™
10936001 |
Phadia™ 5000/2500 | 50 mL | IgE/ECP/Tryptase sample diluent |
Pack of 6 for $111.11
|
N/A | |||
Due to product restrictions, please Sign In to purchase or view availability for this product. | |||||||||
10-931-602P
|
Thermo Scientific™
10931602 |
Phadia™ 250 | 6 x 5.3 mL | Specific IgE Conjugate 100 |
Pack of 600 for $1,602.00
|
|
|||
10-931-002P
|
Thermo Scientific™
10931002 |
Phadia™ 1000 | 6 x 20.5 mL | Specific IgE Conjugate 400 |
Pack of 2400 for $6,408.00
|
|
|||
Description
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title