Learn More
Siemens Healthineers™ Immulite™ 2000 XPI Immunoassay System
Providing multiple tests on a single, easy-to-use analyzer - including allergy and a wide range of specialty offerings - the Immulite™ 2000 XPI Immunoassay System makes it simple to scale your capabilities without disrupting regular day-to-day operations.
Supplier: Siemens Healthineers™ 10373214
This item is not returnable.
View return policy
Description
- Highly reliable Immulite™ 2000 XPI system is easy to use and has one of the largest automated immunoassay menus available
- Medium- to high-volume laboratories can incorporate specialty and allergy testing into routine workloads to improve productivity and efficiency
- Scaling your allergy and specialty test offerings feels routine
- No pause sampling
- 90 day onboard stability to minimize reagent waste
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Analyzer | |
200 Tests/hr. |
Safety and Handling
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov