Learn More
Cargille Immersion Oil
Greatly increases light-gathering power with high-power objectives
Supplier: Cargille 16485/1OZ/EMD
Description
Type NVH is a high-viscosity oil that meets requirements for horizontal, inverted and inclined instruments and projection equipment
- Formulated to give refractive index of 1.5150 ± 0.0002 for D line at 23°C (73°F)
- Nondrying, so index will not change when exposed to air
- Contains no polychlorinated biphenyl (PCB) compounds
- Conforms to ISO 8036/1 specifications
- Standard for all oil-immersion microscopy
Specifications
Immersion Oil Type NVH | |
ISO 8036/1 | |
High-viscosity oil that meets requirements for horizontal, inverted and inclined instruments and projection equipment | |
1.5150 ±0.0002 |
21,000 cSt | |
0.919g/cm3 | |
1 oz. |