Learn More
Cole-Parmer George Ulanet 324-230 Heat-O-Matic Immersion Heater, short Rod , 5" L Heated Area, 1100 watts, 115 VAC
Supplier:  Cole-Parmer 0304604
Encompass_Preferred
Heaters with Dial Temperature Controller have a sensitivity of ±1°>F when liquid is stirred. Top-mounted pilot light indicates that heater is on. Choose from four styles "A" loop, "B" short rod, "C" flat, and "D" long rod which is the same as style "B" only longer. Heaters feature a plastic grip handle; and a 6-ft, three-wire cord, 115 VAC models have U.S. standard plug, 230 VAC models do not have a plug.
Catalog No. NC1569192