Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Agilent BioTek Imaging System Controller, High Performance
This high performance Imaging System Controller is designed for intensive imaging and analysis operations.
$9096.05
Specifications
For Use With (Application) | Intensive Imaging and Analysis |
---|---|
Color | Black |
Includes | Keyboard (1), Mouse (1), Front USB 3.1 Port (4), Rear USB 3.1 Port (6), Mini DisplayPort (3), 1 GB LAN Port (2) |
Display | 24 in. LED Monitor |
Operating System | Microsoft Windows 10 Pro 64-bit |
Catalog Number | Mfr. No. | PC | Includes | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | PC | Includes | Price | Quantity & Availability | ||||
BT1830008
|
Agilent BioTek
1830008S |
Z4 G4 Custom Workstation | Keyboard (1), Mouse (1), Front USB 3.1 Port (4), Rear USB 3.1 Port (6), Mini DisplayPort (3), 1 GB LAN Port (2) |
Each of 1 for $9,096.05
|
|
||||
Description
- Microsoft Windows 10 Pro 64-bit Operating System
- Intel Core i9-7920X Processor (12 core, 2.9 GHz, 16.5 MB cache)
- Adobe Acrobat Reader DC Pre-installed
- Ideal for applications such as:
- Large montages, including single-sample applications
- Complex image analysis on microplate-based assays
- High-object count applications like spot counting
- Higher-throughput imaging application using an Agilent BioTek BioStack microplate stacker or an Agilent BioTek BioSpa automated incubator
Specifications
Intensive Imaging and Analysis | |
Keyboard (1), Mouse (1), Front USB 3.1 Port (4), Rear USB 3.1 Port (6), Mini DisplayPort (3), 1 GB LAN Port (2) | |
Microsoft Windows 10 Pro 64-bit |
Black | |
24 in. LED Monitor | |
Z4 G4 Custom Workstation |
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title