Learn More
PBL Assay Science iLite Human Type I Interferon (IFN) Responsive Cells [2.5 mL/vial]
SDP
Supplier:  PBL Assay Science 511007
Encompass
iLite Human Type I Interferon Responsive Cells are sold by PBL Assay Science (Catalog #51100-7) for research use only (RUO) and packaged in 2.5 mL/vial. These engineered cells are provided as assay ready frozen cells. The cell line is highly specific for its target due to cell engineering using a target-specific reporter gene construct.
Catalog No. 50-153-8076