Learn More
Electron Microscopy Sciences Ilford Ilfospeed RC Deluxe Paper, Glossy-Grade 2
SDP
Supplier:  Electron Microscopy Sciences 8303225
Encompass_Preferred
This is a direct replacement for Kodak Kodabrome II RC B&W Papers. Premium Quality Graded Paper on a Resin Coated Base. MULTIGRADE RC COOLTONE Paper has the standard weight of (190g/m 2) with a resin coated base. It is suitable for a wide range of applications including commercial, press, industrial, advertising, and display work.
Grade: Glossy-Grade 2
Catalog No. 50-313-68