Learn More
MilliporeSigma™ IL-6 ELISPOT Antibody Pair, Human
Protein Interaction Antibody Pairs
Supplier: MilliporeSigma™ ELI008H
Description
Includes:
- Human IL-6 Capture Antibody, clone MQ2-13A5. Quantity: 500 μg.
- Human IL-6 Detection Antibody, clone MQ2-39C3. Quantity: 100 μg.
- Streptavidin Alkaline Phosphatase. Quantity: 55 μL.
Specifications
Monoclonal | |
Rat | |
Primary | |
2-8°C for up to six months | |
Purified |
1.0mg/mL (Capture antibody), 0.5mg/mL (Biotinylated detection antibody) | |
96-well Plates | |
Human | |
MQ2-13A5 (Capture), MQ2-39C3 (Detection) |