Learn More
MilliporeSigma™ IL-4 ELISPOT Antibody Pair, Mouse
Protein Interaction Antibody Pairs
Supplier: MilliporeSigma™ ELI004M
Description
Includes:
- Mouse IL-4 Capture Antibody, clone 11B11. Quantity: 500 μg
- Mouse IL-4 Detection Antibody, clone BVD6-24G2. Quantity: 100 μg.
- Streptavidin Alkaline Phosphatase. Quantity: 55 μL.
Specifications
Monoclonal | |
Rat | |
Primary | |
2-8°C for up to six months | |
Purified |
1.0mg/mL (Capture antibody), 0.5mg/mL (Biotinylated detection antibody) | |
96-well Plates | |
Mouse | |
11B11 (Capture), BVD6 (Detection) |