Learn More
MilliporeSigma™ IL-2 ELISPOT Antibody Pair, Mouse
Protein Interaction Antibody Pairs
Supplier: MilliporeSigma™ ELI002M
Description
Includes:
- Mouse IL-2 Capture Antibody, clone JES6-1A12. Quantity: 500 μ;g.
- Mouse IL-2 Detection Antibody, clone JES6-5H4. Quantity: 100 μg.
- Streptavidin Alkaline Phosphatase. Quantity: 55 μL.
Specifications
Monoclonal | |
Rat | |
Primary | |
2-8°C for up to six months | |
Purified |
1.0mg/mL (Capture antibody), 0.5mg/mL (Biotinylated detection antibody) | |
96-well Plates | |
Mouse | |
JES6-1A12 (Capture), JES6-5H4 (Detection) |