Learn More
MilliporeSigma™ IL-10 ELISPOT Antibody Pair, Human
Protein Interaction Antibody Pairs
Supplier: MilliporeSigma™ ELI010H
Description
Includes:
- Human IL-10 Capture Antibody, clone JES3-19F1. Quantity: 500 μg.
- Human IL-10 Detection Antibody, clone JES3-12G8. Quantity: 100 μg.
- Streptavidin Alkaline Phosphatase. Quantity: 55 μL.
Specifications
Monoclonal | |
Rat | |
Primary | |
2-8°C for up to six months | |
Purified |
1.0mg/mL (Capture antibody), 0.5mg/mL (Biotinylated detection antibody) | |
96-well Plates | |
Human | |
JES3-19F1 (Capture), JES3-12G8 (Detection) |