Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IgG1, κ, Mouse, Alexa Fluor™ 647, Clone: MOPC-31C; Isotype Control, BD Pharmingen™
Mouse Monoclonal Isotype Control
Supplier: BD Biosciences 566011
Description
IgG1, κ Monoclonal antibody specifically detects IgG1, κ in samples. It is validated for Flow cytometry (Intracellular Staining)), Isotype control.Specifications
IgG1 κ | |
Monoclonal | |
0.2mg/mL | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
50 μg | |
Cell Biology | |
Purified |
Flow Cytometry | |
MOPC-31C | |
Alexa Fluor 647 | |
Mouse | |
RUO | |
Primary | |
IgG1 κ |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
For Reseach Use Only.
Spot an opportunity for improvement?
Provide Content Correction