Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IgG1 Kappa Human, PE, Isotype Control, Novus Biologicals™
Human Polyclonal Isotype Control
Supplier: Novus Biologicals NBP306872PE
This item is not returnable.
View return policy
Description
IgG1 Kappa Polyclonal antibody specifically detects IgG1 Kappa in Human samples. It is validated for ControlSpecifications
IgG1 Kappa | |
PE | |
Human | |
0.1 mL | |
Human | |
Isotype Controls |
Polyclonal | |
PBS | |
Human myeloma serum | |
RUO | |
Store at 4°C in the dark. | |
IgG1 κ |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction