Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IgG Fc Sheep anti-Guinea Pig, FITC, Polyclonal, Secondary Antibody, Novus Biologicals™
Sheep Polyclonal Antibody
Supplier: Novus Biologicals NBP269480
Description
IgG Fc Polyclonal antibody specifically detects IgG Fc in Guinea Pig samples. It is validated for ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Frozen).Specifications
IgG Fc | |
Polyclonal | |
FITC | |
Lyophilized from PBS (pH 7.2) with No Preservative | |
Purified normal IgG isolated from pooled guinea pig serum. Freund's complete adjuvant is used in the first step of the immunization procedure. | |
1 mL | |
The reactivity of the antiserum is directed to the Fc subunit of the IgG molecule. In immunoelectrophoresis and double radial immunodiffusion, using various antiserum concentrations against normal guinea pig plasma and serum, a single characteristic precipitin line is obtained which shows a reaction of identity with the precipitin line obtained with purified IgG. No reaction is obtained with purified IgA, IgM and IgG/Fab fragments. | |
Guinea Pig | |
Lyophilized |
ELISA, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Frozen) | |
LYOPH | |
ELISA 1:100-1:2000, Immunocytochemistry/Immunofluorescence 1:10 - 1:500, Immunohistochemistry-Frozen 1:10 - 1:500 | |
Sheep | |
Ion exchange chromatography | |
Secondary | |
Reconstitute by by adding 1 ml sterile distilled water, spun down to remove insoluble particles, divided into small aliquots, frozen and stored at or below -20C. | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction