Learn More
Advanced ImmunoChemical, Inc. Recombinant Human Insulin-Like Growth Factor Binding Protein 5 (IGFBP-5), Source - Expressed In Mammalian Cell Line Full Length Mature Protein, Analysis:Purity > 90% (According To SDS-PAAG), Including Detected Fragments, 0.05mg
SDP
Supplier:  Advanced ImmunoChemical, Inc. 8IGBP50.05MG
Encompass_Preferred
This item is manufactured by HyTest, Ltd and distributed by Advanced ImmunoChemical, Inc. Recombinant human Insulin-like growth factor binding protein 5 (IGFBP-5). Source: Expressed in mammalian cell line. Description: IGFBP-5 is expressed as full length mature protein (amino acids 21-272, UniProt ID = P24593). Applications: Suitable for use as a mass IGFBP-5 standard, standard or calibrator in immunoassay. Analysis: Purity > 90% (according to SDS-PAAG), including detected fragments. IGFBP-5 concentration is determined spectrophotometrically. Immunoreactivity is confirmed in sandwich immunoassay with specific monoclonal antibodies. Purification: Immunoaffinity purification. Presentation:40 mM sodium citrate pH 4.0Storage Conditions:-70 °C (between -65 and -80 °C allowed). Other information: Avoid repeated freezing and thawing. It is recommended to aliquot the product after the first thawing. Store the protein at pH 4.0.
Catalog No. 50-153-9841