Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Ig, κ light chain Rat anti-Mouse, RB780, Clone: 187.1, BD OptiBuild™
Rat Monoclonal Antibody
Supplier: BD Biosciences 755937
This item is not returnable.
View return policy
Description
Ig, κ light chain Monoclonal antibody specifically detects Ig, κ light chain in Mouse samples. It is validated for Flow CytometrySpecifications
Ig, κ light chain | |
Monoclonal | |
0.2 mg/mL | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
Rat | |
50 μg | |
Primary | |
Mouse | |
IgG1 κ |
Flow Cytometry | |
187.1 | |
RB780 | |
Ig kappa chain C region; IGKC; Igk-C; Ig, κ | |
Mouse IgG2b κ secreted by MPC-11 plasmacytoma | |
RUO | |
16071 | |
Store undiluted at 4°C and protected from prolonged exposure to light.Do not freeze.The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography.The antibody was conjugated to the dye under optimum conditions that minimize unconjugated dye and antibody. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction