Learn More
Cole-Parmer FSB-200-I-5L Industrial Fluidized Sand Bath, 5 L
Supplier: Cole-Parmer 0118410
Industrial sand baths are ideal for heat treatment, cleaning and temperature calibrations, as well as reactive analysis, such as exothermic and endothermic reactions. They remove most polymers from tooling, including PVC, PET, PTFE, and PEEK without abrasion to the tool or mold. Parts immersed into the bath are cleaned by the high temperature -- up to 600C /1112F environment within a bath media of fluidized aluminum oxide. Baskets are available for cleaning of small parts. All baths require a dry air supply or an inert gas, such as nitrogen and argon. Each unit comes with a front panel flowmeter to adjust airflow with settings for optimal flow per temperature. When cleaning parts, baths should be operated in a hood or a well-ventilated area. Extraction collar and fans are available to ensure proper removal of fumes.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.