Learn More
Medchemexpress LLC HY-101479 5mg Medchemexpress, Iclaprim CAS:192314-93-5 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1014795MG
Encompass_Preferred
Medchemexpress, HY-101479 5mg Iclaprim CAS:192314-93-5 Iclaprim is a new selective bacterial Dihydrofolate inhibitor, which can inhibit the growth of S. aureus ( MRSA ) with an MIC 90 of 0.06 μg/mL. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-2145