Learn More
ProteinSimple ICE280 CHEMICAL TEST KIT
Supplier: ProteinSimple 101801
![Encompass](/content/dam/fishersci/glyphs/encompass.png)
The Chemical Test Kit is designed to confirm the overall performance of the iCE3 or iCE280 with the FC cIEF cartridge on 3 separate occasions. Each time, performance can be checked repeatedly over a 3 day period. FC Cartridge Chemical Test Kit. This kit includes anolyte, catholyte, 0.5% methyl cellulose, a hemoglobin stock solution and three vials of a standard solution (carrier ampholytes and pI markers in 0.35% methyl cellulose).
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.