Learn More
ProteinSimple iCE Electrolyte Kit 2 x 100 milliliters
Supplier: ProteinSimple 102506
These 100mL Anolyte and Catholyte solutions in 0.1% MC are used to fill the electrolyte tanks on both the FC and HT cIEF cartridges. The labels and bottles are color coded to improve safety. The kit contains 2 x 100 mL bottles.