Learn More
Electron Microscopy Sciences EMS Model IAM-MET, ASTM Grain Size, 32 pts Calibrated
Supplier: Electron Microscopy Sciences 60216METC
The ASTM Grain Size / Nodularity Analysis Standard is based on the IAM-1 Morphometric Image Analysis above, the IAM-MET is a multi-morphology calibration standard designed specifically for ASTM. The IAM-MET includes all plates as found in IAM-1 except for plate 4. This plate contains 9 images of irregular shapes, simulating features commonly found in metallurgical applications. As required by the ASTM standard, the IAM-MET includes secondary certification of plates 1, 2 and 8. Calibrated.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.