Learn More
AVANTOR MASTERFLEX (VWR LLC) Masterflex I/P Digital Modular Drive with Benchtop Controller and Remote Input/Output, 0.1 to 650 rpm; 115/230 VAC
Supplier:  AVANTOR MASTERFLEX (VWR LLC) MFLX0759400
Encompass_Preferred
These I/P digital pumps give you rugged performance and precise digital control. These full featured drives are ideal for fluid transfer, dispensing, and metering applications in laboratory, process, and industrial environments including pharmaceutical, food & beverage, dairy, chemical, manufacturing, and water treatment. Precision drives feature a programmable dispense interval that lets you set the delay between dispense cycles for convenient, automated dispensing by time or by volume. Programmed calibration ensures dispense and display accuracy¿drive stores one calibration value per tubing size. Program the drive via the simple membrane keypad; keypad lock/unlock feature protects settings. Select tubing size and enter desired flow rate¿drive locks in required rpm.
Catalog No. NC0675190