Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mirus Bio™ flashBAC™ ULTRA Baculovirus Expression System

flashBAC™ ULTRA Baculovirus Expression System is a streamlined platform for the production of recombinant baculoviruses. Ideal for the most difficult to express proteins including nuclear, cytoplasmic, membrane and secreted proteins.

Specifications

Content And Storage Store flashBAC™ DNA at 4°C and control plasmid at −20°C
Cell Type Insect Cells
Format Kit
Sample Type Baculovirus DNA, Plasmid DNA
Transfection Technique Chemical Transfection
View More Specs

Products 2
Catalog Number Mfr. No. No. of Reactions Price Quantity & Availability  
Catalog Number Mfr. No. No. of Reactions Price Quantity & Availability  
MIR6135
View Documents Promotion Details
Mirus Bio™
MIR 6135
5
Each for $1,437.00
N/A
Due to product restrictions, please to purchase or view availability for this product.
MIR6140
View Documents Promotion Details
Mirus Bio™
MIR 6140
24
Each for $3,849.00
N/A
Due to product restrictions, please to purchase or view availability for this product.
Description

Description

flashBAC expression systems are a streamlined platform for the production of recombinant baculoviruses. Genetic optimization of the Autographa californica nucleopolyhedrovirus (AcMNPV) genome yields recombinant virus in a single step and eliminates the need for separation of recombinant and parental virus by plaque purification. flashBAC ULTRA is the most advanced flashBAC vector system, optimized by deletions in the chitinase, cathepsin, p10, p74 and p26 genes. The ideal system for the most difficult to express proteins including nuclear, cytoplasmic, membrane and secreted proteins.
  • Single-step baculovirus generation in insect cells, no plaque purification required
  • Ready to use with most transfer vectors, including BacMam
  • High recombinant protein yields

Warranty and Services

1 Year

Specifications

Specifications

Store flashBAC™ DNA at 4°C and control plasmid at −20°C
Kit
Chemical Transfection
Sterile
Insect Cells
Baculovirus DNA, Plasmid DNA
Transfection
SDS
Documents

Documents

Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit