Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Abnova™ E. Coli O157:H7 ELISA Kit
Sandwich enzyme immunoassay for the quantitative measurement of E. Coli O157:H7
Supplier: Abnova™ KA4787
Description
- For quantitative detection of E.coli O157:H7 in food or water
Specifications
6.25 × 103 to 4 ×105 cfu/mL | |
HRP | |
ELISA Kit | |
E. coli | |
One 96-well plate pre-coated with anti-E.coli O157:H7 antibody, Lyophilized E.coli O157:H7 standards: 4 × 105 cfu / tube, Sample / diluent buffer, HRP conjugated anti-E.coli O157:H7 antibody (Concentrated): Dilution: 1:100, Antibody diluent buffer, TMB substrate, Stop solution, Wash buffer: (25x): Dilution: 1:25 | |
100 μL |
E. Coli O157:H7 ELISA Kit is a sandwich enzyme immunoassay for the quantitative measurement of E. Coli O157:H7. | |
Colorimetric | |
Food, Water | |
Quantification | |
For research use only (RUO) | |
Store at 4°C for 4 months, or at -20°C for 8 months. Store HRP conjugated antibody at 2-8°C. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction