Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Hypromellose, Substitution Type 2208, 100 mPa.s, USP, 4-12%, Spectrum™ Chemical
Click to view available options
Quantity:
125 g
500 g
2.5 kg
12 kg
45 kg
Packaging:
Fiber Drum
Poly Bottle
Poly Pail
Description
Spectrum™ Chemical Hypromellose, Substitution Type 2208, 100 mPa.s, USP is a semisynthetic, viscoelastic polymer that is inert and used as an ophthalmic lubricant, a controlled-delivery component and an excipient in oral medicaments. It is also used as a food additive and acts as an emulsifier, suspending agent and thickening agent. Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
Specifications
CAS Max % | 100% |
Quantity | 45 kg |
Percent Purity | 4 to 12% |
Grade | USP |
Ignition Residue | 1.5% |
Loss on Drying | 5% |
Packaging | Fiber Drum |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction